ENA156508250 Enamine

3-[(methoxyacetyl)amino]propanoic acid

  • MDL number MFCD09050610
  • Cas Number : 36855-37-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31242

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31242
Availability details for
Quantity
  • Availability
  • Add to Cart