ENA156508251 Enamine

N-phenyl-1,3-thiazolidine-4-carboxamide hydrochloride

  • MDL number MFCD09971432
  • Cas Number : 1078162-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-31243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31243
Availability details for
Quantity
  • Availability
  • Add to Cart