ENA156508259 Enamine

3-(phenylamino)-1,2-dihydroisoquinolin-1-one

  • MDL number MFCD06405085
  • Cas Number : 56100-50-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31255

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31255
Availability details for
Quantity
  • Availability
  • Add to Cart