ENA156508262 Enamine

(4,6-dimethylpyrimidin-2-yl)acetonitrile

  • MDL number MFCD09971438
  • Cas Number : 32691-58-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31258
Availability details for
Quantity
  • Availability
  • Add to Cart