ENA156508268 Enamine

5-ethyl-1-phenyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971441
  • Cas Number : 116344-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31265
Availability details for
Quantity
  • Availability
  • Add to Cart