ENA156508269 Enamine

N-[1-(2,4-dichlorophenyl)ethyl]-N-methylamine

  • MDL number MFCD09734142
  • Cas Number : 953898-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31266

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31266
Availability details for
Quantity
  • Availability
  • Add to Cart