ENA156508274 Enamine

N-(3-methylbutyl)cyclopropanamine hydrochloride

  • MDL number MFCD09971442
  • Cas Number : 1093403-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31271

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-31271
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart