ENA156508277 Enamine

7-(trifluoromethyl)pyrido[2,3-d]pyrimidine-2,4(1H,3H)-dione

  • MDL number MFCD09971445
  • Cas Number : 779352-04-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31276

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31276
Availability details for
Quantity
  • Availability
  • Add to Cart