ENA156508278 Enamine

3-[4-(cyclopropylsulfamoyl)phenyl]propanoic acid

  • MDL number MFCD08445723
  • Cas Number : 889939-54-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31278

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31278
Availability details for
Quantity
  • Availability
  • Add to Cart