ENA156508291 Enamine

[2-(morpholin-4-yl)-4-phenyl-1,3-thiazol-5-yl]methanol

  • MDL number MFCD08442501
  • Cas Number : 926228-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31296
Availability details for
Quantity
  • Availability
  • Add to Cart