ENA156508293 Enamine

3-[(4-fluorophenyl)methoxy]aniline

  • MDL number MFCD07801209
  • Cas Number : 79808-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31298

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31298
Availability details for
Quantity
  • Availability
  • Add to Cart