ENA156508324 Enamine

methyl 4-chloro-3-(chlorosulfonyl)benzoate

  • MDL number MFCD09816501
  • Cas Number : 1000933-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-157
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31348

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31348
Availability details for
Quantity
  • Availability
  • Add to Cart