ENA156508325 Enamine

2-chloro-5-methanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD08445060
  • Cas Number : 90084-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31349

Safety Information

signalword Warning
pcodes P264,P280,P305+P351+P338,P337+P313
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-31349
pictograms GHS07
hstatements H319
Availability details for
Quantity
  • Availability
  • Add to Cart