ENA156508339 Enamine

3-(2,5-dioxopyrrolidin-1-yl)benzonitrile

  • MDL number MFCD00176070
  • Cas Number : 72601-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 134...136
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31370

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31370
Availability details for
Quantity
  • Availability
  • Add to Cart