ENA156508342 Enamine

4-ethyl-3-(morpholine-4-sulfonyl)benzoic acid

  • MDL number MFCD09050946
  • Cas Number : 926232-25-9
  • Molecular Weight : 299.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31373
Availability details for
Quantity
  • Availability
  • Add to Cart