ENA156508343 Enamine

3-(azetidine-1-sulfonyl)-4-fluorobenzoic acid

  • MDL number MFCD09040968
  • Cas Number : 926259-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31374
Availability details for
Quantity
  • Availability
  • Add to Cart