ENA156508345 Enamine

1-chloro-4-methylphthalazine

  • MDL number MFCD00631939
  • Cas Number : 19064-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123...125
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18472

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18472
Availability details for
Quantity
  • Availability
  • Add to Cart