ENA156508346 Enamine

N-(3-cyanophenyl)-2-methoxyacetamide

  • MDL number MFCD07751787
  • Cas Number : 900711-77-5
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31377
Availability details for
Quantity
  • Availability
  • Add to Cart