ENA156508349 Enamine

3-(2,4-dimethylphenoxy)propanoic acid

  • MDL number MFCD09044182
  • Cas Number : 15904-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31380

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31380
Availability details for
Quantity
  • Availability
  • Add to Cart