ENA156508359 Enamine

3-[3-(4-ethoxyphenyl)-1,2,4-oxadiazol-5-yl]propanoic acid

  • MDL number MFCD05130926
  • Cas Number : 500026-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31399

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31399
Availability details for
Quantity
  • Availability
  • Add to Cart