ENA156508363 Enamine

N-(3-cyanophenyl)cyclobutanecarboxamide

  • MDL number MFCD03948386
  • Cas Number : 750620-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31421

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31421
Availability details for
Quantity
  • Availability
  • Add to Cart