ENA156508366 Enamine

5-ethyl-1-(4-methylphenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971486
  • Cas Number : 1002185-78-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31424

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31424
Availability details for
Quantity
  • Availability
  • Add to Cart