ENA156508367 Enamine

1-cyclopropyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbonitrile

  • MDL number MFCD00203325
  • Cas Number : 175203-44-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31425
Availability details for
Quantity
  • Availability
  • Add to Cart