ENA156508374 Enamine

1-(2,3-dihydro-1H-inden-5-yl)-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD08445224
  • Cas Number : 923106-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31433

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31433
Availability details for
Quantity
  • Availability
  • Add to Cart