ENA156508385 Enamine

1-methyl-2,4-dioxo-7-phenyl-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD08436277
  • Cas Number : 889941-16-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31473
Availability details for
Quantity
  • Availability
  • Add to Cart