ENA156508400 Enamine

1-(3-chlorophenyl)-5-(propan-2-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD09971499
  • Cas Number : 1004043-11-1
  • Molecular Weight : 264.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31491

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31491
Availability details for
Quantity
  • Availability
  • Add to Cart