ENA156508402 Enamine

1-[2-(morpholin-4-yl)ethyl]-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09064345
  • Cas Number : 304859-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-30978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30978
Availability details for
Quantity
  • Availability
  • Add to Cart