ENA156508403 Enamine

1-(butane-1-sulfonyl)piperidine-2-carboxylic acid

  • MDL number MFCD08442560
  • Cas Number : 1105682-81-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31496
Availability details for
Quantity
  • Availability
  • Add to Cart