ENA156508405 Enamine

N-(4-bromophenyl)pyrrolidine-2-carboxamide hydrochloride

  • MDL number MFCD09863853
  • Cas Number : 1078162-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30980

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30980
Availability details for
Quantity
  • Availability
  • Add to Cart