ENA156508409 Enamine

4,7-dioxo-7-phenylheptanoic acid

  • MDL number MFCD00020544
  • Cas Number : 6336-53-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31507

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31507
Availability details for
Quantity
  • Availability
  • Add to Cart