ENA156508417 Enamine

3-(naphthalen-1-yloxy)propanoic acid

  • MDL number MFCD00156915
  • Cas Number : 16563-41-0
  • Molecular Weight : 216.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 142-143
Iupac Name 3-(1-naphthyloxy)propanoic acid
Inchi Code 1S/C13H12O3/c14-13(15)8-9-16-12-7-3-5-10-4-1-2-6-11(10)12/h1-7H,8-9H2,(H,14,15)
Inchi Key JPLBWCCELAOKOU-UHFFFAOYSA-N

Description

Melting Point 142-143
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-31521

Safety Information

signalword Warning
pcodes P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-31521
pictograms GHS07
hstatements H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart