ENA156508431 Enamine

2-[(2-amino-4-methyl-1,3-thiazol-5-yl)sulfanyl]acetamide

  • MDL number MFCD09935034
  • Cas Number : 1000933-46-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31541

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31541
Availability details for
Quantity
  • Availability
  • Add to Cart