ENA156508432 Enamine

1-(2-cyanoacetyl)piperidine-4-carboxamide

  • MDL number MFCD02373746
  • Cas Number : 328384-63-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-30999

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30999
Availability details for
Quantity
  • Availability
  • Add to Cart