ENA156508434 Enamine

ethyl 1-(chlorosulfonyl)pyrrolidine-2-carboxylate

  • MDL number MFCD08445192
  • Cas Number : 1028366-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31545

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31545
Availability details for
Quantity
  • Availability
  • Add to Cart