ENA156508435 Enamine

N-(3-amino-4-fluorophenyl)-1,3-dioxaindane-5-carboxamide

  • MDL number MFCD09048198
  • Cas Number : 926265-21-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-31548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31548
Availability details for
Quantity
  • Availability
  • Add to Cart