ENA156508438 Enamine

5-amino-4-methyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD09971375
  • Cas Number : 66870-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 268-270
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31000

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31000
Availability details for
Quantity
  • Availability
  • Add to Cart