ENA156508451 Enamine

1-[4-(trifluoromethyl)benzoyl]piperazine hydrochloride

  • MDL number MFCD09971519
  • Cas Number : 1171417-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31566
Availability details for
Quantity
  • Availability
  • Add to Cart