ENA156508456 Enamine

ethyl 1-ethyl-2-oxo-2,3-dihydro-1H-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD09971378
  • Cas Number : 1000932-51-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31014

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31014
Availability details for
Quantity
  • Availability
  • Add to Cart