ENA156508459 Enamine

tert-butyl[(2,5-dimethoxyphenyl)methyl]amine

  • MDL number MFCD09735550
  • Cas Number : 953891-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31579
Availability details for
Quantity
  • Availability
  • Add to Cart