ENA156508460 Enamine

N-[(2,5-dimethoxyphenyl)methyl]cyclopropanamine

  • MDL number MFCD09740511
  • Cas Number : 877996-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31580
Availability details for
Quantity
  • Availability
  • Add to Cart