ENA156508462 Enamine

N-(2-amino-5-chlorophenyl)furan-2-carboxamide

  • MDL number MFCD09050720
  • Cas Number : 926197-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31583
Availability details for
Quantity
  • Availability
  • Add to Cart