ENA156508466 Enamine

2-[4-(1-hydroxyethyl)phenoxy]-N-methylacetamide

  • MDL number MFCD08442730
  • Cas Number : 926185-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31593

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31593
Availability details for
Quantity
  • Availability
  • Add to Cart