ENA156508469 Enamine

5-(3-bromo-4-fluorophenyl)imidazolidine-2,4-dione

  • MDL number MFCD09731233
  • Cas Number : 1046118-65-3
  • Molecular Weight : 273.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31604
Availability details for
Quantity
  • Availability
  • Add to Cart