ENA156508474 Enamine

2-(methoxymethyl)benzonitrile

  • MDL number MFCD09944124
  • Cas Number : 57991-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31633

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31633
Availability details for
Quantity
  • Availability
  • Add to Cart