ENA156508476 Enamine

2-(3-methoxyphenoxy)ethanamine hydrochloride

  • MDL number MFCD00235191
  • Cas Number : 26378-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140...142
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31636

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31636
Availability details for
Quantity
  • Availability
  • Add to Cart