ENA156508478 Enamine

2-(4-ethoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD06857932
  • Cas Number : 58765-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-31644

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-31644
Availability details for
Quantity
  • Availability
  • Add to Cart