ENA156508480 Enamine

N-{3-[(aminocarbonothioyl)amino]phenyl}acetamide

  • MDL number MFCD09971530
  • Cas Number : 1614-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31648

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31648
Availability details for
Quantity
  • Availability
  • Add to Cart