ENA156508481 Enamine

2-methyl-3-nitrobenzenesulfonyl chloride

  • MDL number MFCD08443419
  • Cas Number : 56682-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 47...49
Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31020

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31020
Availability details for
Quantity
  • Availability
  • Add to Cart