ENA156508498 Enamine

1H-imidazol-2-ylacetonitrile

  • MDL number MFCD08668414
  • Cas Number : 23184-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31693

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31693
Availability details for
Quantity
  • Availability
  • Add to Cart