ENA156508503 Enamine

1-(3-chloropropanoyl)-4-(4-methoxyphenyl)piperazine

  • MDL number MFCD09809199
  • Cas Number : 1000932-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-31036

Safety Information

msds https://www.enaminestore.com/catalog/EN300-31036
Availability details for
Quantity
  • Availability
  • Add to Cart